Learn More
Thermo Scientific™ Universal Microplate Lids
GREENER_CHOICE
Thermo Scientific™ Universal Microplate Lids fit all microplates including Thermo Scientific™ Microtiter™ Immulon™ microplates.
Brand:  Thermo Scientific™ 5500
2609.63 NOK valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Product Code. 11912627
  • 3280.00 NOK / Pack of 100
Only null left
In Stock
Description
Thermo Scientific™ Universal Microplate Lids fit all microplates including Thermo Scientific™ Microtiter™ Immulon™ microplates.
  • Universal
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
100
Cover
96
50
All Microplates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5417169 Certificate TypeCertificate of Processing Date11/06/2025 Catalog Number5500
Lot Number5412778 Certificate TypeCertificate of Processing Date12/05/2025 Catalog Number5500
Lot Number5394312 Certificate TypeCertificate of Processing Date12/05/2025 Catalog Number5500
Lot Number5394312 Certificate TypeCertificate of Compliance Date16/05/2025 Catalog Number5500
Lot Number5412778 Certificate TypeCertificate of Compliance Date16/05/2025 Catalog Number5500
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Universal Microplate Lids > Sterile