Learn More
Usbeck Laborgeraete™ Steel Triangles
Brand:  Usbeck Laborgeraete™ 5104
 View more versions of this product
Product Code. 10433121
  • 46.40 NOK / Each
Only null left
Estimated Shipment: 07-07-2025
Specifications
1 Each
80 mm
Triangle
Triangles
Steel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Steel Triangles > Length: 80mm