Learn More
Usbeck Laborgeraete™ Steel Triangles
Brand:  Usbeck Laborgeraete™ 5100
 View more versions of this product
Product Code. 10303101
  • 38.65 NOK / Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
1 Each
40 mm
Triangle
Triangles
Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Steel Triangles > Length: 40mm