Learn More
Usbeck Laborgeraete™ Steel Triangles
Brand:  Usbeck Laborgeraete™ 5101
 View more versions of this product
Product Code. 10189740
  • 40.00 NOK / Each
Only null left
Estimated Shipment: 07-07-2025
Specifications
1 Each
50 mm
Triangle
Triangles
Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Steel Triangles > Length: 50mm