Learn More
Fisherbrand™ Stainless Steel Sharp Ends Forceps
Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Sharp Ends Forceps provide excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized.
213.00 NOK - 251.00 NOK
Specifications
Tip Style | Sharp |
---|---|
No. per Pack | Each |
Material | PTFE-coated Stainless Steel |
Product Type | Forceps |
Style | Serrated Jaws |
Product Code | Brand | Height (Metric) Body | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) Body | Price | Quantity & Availability | ||||||
11562113
|
Fisherbrand™
7201 |
115 mm |
213.00 NOK
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
11572113
|
Fisherbrand™
7202 |
130 mm |
251.00 NOK
Each |
In Stock
Log in to see stock. ![]() |
||||||
11582113
|
Fisherbrand™
7204 |
160 mm |
248.00 NOK
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.- Chemically resistant and non-contaminating
- Non-protein binding and providing bio-inert surfaces
- No critical quantities lost due to sticking
- No build up on surfaces
- Ideal for use under severe laboratory conditions
- Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
- With guide pin
Specifications
Sharp | |
PTFE-coated Stainless Steel | |
Serrated Jaws |
Each | |
Forceps |
Product Title
Fisherbrand™ Stainless Steel Sharp Ends Forceps