Learn More
Bochem™ Stainless Steel Laboratory Scissors with Stopper Lifter
Manufactured from stainless steel
Brand:  Bochem™ 4200
Product Code. 10547931
  • 111.00 NOK / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Description
  • Tip style: sharp-round
  • Stainless steel
  • With stopper lifter
Specifications
Specifications
Laboratory Scissors
150 mm
Stainless Steel
Sharp/Round
Documents
One moment while we fetch your results.
Product Title
Bochem™ Stainless Steel Laboratory Scissors with Stopper Lifter > Length/Tip Style: 150mm/Sharp/Round