Learn More
Bochem™ Stainless Steel Tissue Forceps
Manufactured from stainless steel. BOCHEM™ Stainless Steel Tissue Forceps serve a variety of laboratry uses.
Brand:  Bochem™ 1153
Product Code. 10098140
  • 60.50 NOK / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Description
  • Stainless steel construction
  • Round tips
Specifications
Specifications
1 Pack
160 mm
Teeth
Forceps, Tissue
Stainless Steel
Forceps
1 x 2
Documents
One moment while we fetch your results.
Product Title
Bochem™ Stainless Steel Tissue Forceps > Length: 160mm