missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™

ACS Reagent,≥99.5%

342.70 NOK - 1998.70 NOK

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrateShow MoreShow Less
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15612050
SDS View Documents
Honeywell-Fluka
217247-25G
25 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15622050
SDS View Documents
Honeywell-Fluka
217247-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15602050
SDS View Documents
Honeywell-Fluka
217247-2.5KG
2.5 kg Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1080 days from date of manufacture

missing translation for 'einecsNumber' : 231-867-5

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.