missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.50kg
25g
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
CAS Max % | 100.0000% |
Quantity | 25 g |
Linear Formula | Na2S2O3 · 5H2O |
UN Number | NONH for all modes of transport |
Solubility Information | Soluble in water |
Formula Weight | 248.18 |
Percent Purity | ≥99.5% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Density | 1.73 g/cm3 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction