missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS Reagent Grade, Scharlab

Used in analytical chemistry, as a laboratory reagent, to make sodium salts, in the food industry and for decreasing the melting point of water.

279.00 NOK - 510.00 NOK

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318544
View Documents
Scharlab
SO02271000
1 kg
279.00 NOK
1kg
Please to purchase this item. Need a web account? Register with us today!
11328544
View Documents
Scharlab
SO0227005P
5 kg
510.00 NOK
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Synonyms: Salt, Common salt, Rock salt, Sea salt
  • Molecular Formula: NaCl
  • Molar Mass: 58.44g/mol
  • CAS: 7647-14-5
  • EIN ECS No.: 231-598-3
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Chloride, ACS Reagent Grade, Scharlab

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.