missing translation for 'onlineSavingsMsg'
Learn More
Sodium Chloride, ACS Reagent Grade, Scharlab Product Code.: 11318544

Sodium Chloride, ACS Reagent Grade, Scharlab

Product Code. 11318544
1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11318544

Brand: Scharlab SO02271000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, to make sodium salts, in the food industry and for decreasing the melting point of water.

  • Synonyms: Salt, Common salt, Rock salt, Sea salt
  • Molecular Formula: NaCl
  • Molar Mass: 58.44g/mol
  • CAS: 7647-14-5
  • EIN ECS No.: 231-598-3

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
pH 5.0–8.0 (5%, water)
Quantity 1 kg
Formula Weight 58.44g/mol
Percent Purity 99.5% min (argentometric)
Grade ACS Reagent
Identification Passes Test
Loss on Drying 0.5% max (105°C, 2h)
Vapor Pressure 1.3 hPa (at 865°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Chloride, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.