missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, ≥99.5%, Reagent Grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
2.50kg
500g
5kg
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Color | Colorless |
pH | 8.0 to 8.6 (at 20°C) |
Quantity | 500 g |
Linear Formula | NaHCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4153970 |
IUPAC Name | sodium;hydrogen carbonate |
Formula Weight | 84.01g/mol |
Percent Purity | ≥99.5% |
Odor | Odorless |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Bicarbonate, ≥99.5%, Reagent Grade, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction