missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydrogen Carbonate, ACS Reagent Grade, Scharlab Product Code.: 11398594

Sodium Hydrogen Carbonate, ACS Reagent Grade, Scharlab

Product Code. 11398594
1 kg, 1kg
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11398594

Brand: Scharlab SO01311000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, to make sodium salts, for laboratory glassware cleaning, as a source of CO2, in fire extinguishers and in the food industry (E-500).

  • Synonyms: Sodium bicarbonate, Baking soda
  • Molecular Formula: NaHCO3
  • Molar Mass: 84.01g/mol
  • CAS: 144-55-8
  • EIN ECS No.: 205-633-8
  • Phosphates and Silicates (as SiO2): max. 0.005%
  • Sulfur Compounds (as SO4): max. 0.003%
  • Substances Reducing Iodine: max. 0.0065%

Specifications

Melting Point 270°C (decomposition)
CAS 144-55-8
pH 8.6% max (5%, water)
Quantity 1 kg
Molecular Formula CHNaO3
MDL Number MFCD00003528
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES [Na+].OC([O-])=O
IUPAC Name sodium hydrogen carbonate
Molecular Weight (g/mol) 84.01
PubChem CID 516892
ChEBI CHEBI:32139
Formula Weight 84.01g/mol
Percent Purity 99.7 to 100.3% (acidimetric, on dried sample)
Grade ACS Reagent
Identification Passes Test
Loss on Drying 0.2% max (silica gel)
Vapor Pressure 8.3hPa (at 30°C)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydrogen Carbonate, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.