missing translation for 'onlineSavingsMsg'
Learn More
Sodium Bicarbonate, ≥99.5%, Reagent Grade, Honeywell™ Product Code.: 15628130

Sodium Bicarbonate, ≥99.5%, Reagent Grade, Honeywell™

Product Code. 15628130 Shop All Honeywell Research Chemicals Products
500 g, Poly bottle, 500g
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
2.50kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15628130

Brand: Honeywell Research Chemicals S8875500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, powder,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow MoreShow Less
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Color Colorless
pH 8.0 to 8.6 (at 20°C)
Quantity 500 g
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly bottle
Appearance Powder
Density 2.2g/cu.cm at 20°C
Chemical Name or Material Sodium bicarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Bicarbonate, ≥99.5%, Reagent Grade, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.