missing translation for 'onlineSavingsMsg'
Learn More
Sodium bicarbonate, 99.5%, MP Biomedicals™ Product Code.: 11415442

Sodium bicarbonate, 99.5%, MP Biomedicals™

Product Code. 11415442 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
This item is not returnable. View return policy

Product Code. 11415442

Brand: MP Biomedicals 0219549701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium bicarbonate ≥99.5%, White Powder

  • It is broadly used as a physiological buffer in cell culture applications.
  • The pKa for bicarbonate <-> carbonic acid reaction is 6.4 The pKa for bicarbonate <-> carbonate reaction is 10.3.
  • It has two pKas, 6.4 and 10.3. The compound can react with both acids and bases, therefore it can be used as a neutralizer.
  • It may be used to remove or wash acidic impurities from a crude liquid to yield a purer sample.
  • it reacts with the carboxyl groups to give a fast-forming and effervescent CO2 formation which has been used to test for the presence of carboxylic groups in proteins.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.5%
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
Formula Weight 84.01
Percent Purity 99.5%
Density 2.159g/mL
Chemical Name or Material Sodium bicarbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium bicarbonate, 99.5%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.