missing translation for 'onlineSavingsMsg'
Learn More
Learn More

Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium Bicarbonate, 99.5%-100.0%, (Crystalline powder/ACS Reagent Grade), MP Biomedicals
Product Code. 11404642 Shop All MP Biomedicals Products1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable.
View return policy
Product Code. 11404642
Brand: MP Biomedicals 0219143501
This item is not returnable.
View return policy
Specifications
Specifications
CAS | 144-55-8 |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Assay Percent Range | 99.5%-100.0% |
Molecular Formula | CHNaO3 |
MDL Number | MFCD00003528 |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
SMILES | [Na+].OC([O-])=O |
IUPAC Name | sodium hydrogen carbonate |
Molecular Weight (g/mol) | 84.01 |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
Formula Weight | 84.01 |
Grade | ACS Reagent |
Chemical Name or Material | Sodium Bicarbonate |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 10118190
-
Product Code. 15641310
-
-
-
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 6
-
Catalog No. 10703811
kr 2 195,00 / 500g -
Catalog No. 10793611
kr 815,00 / 1kg -
Catalog No. 10117113
kr 444,42 / 100g -
Catalog No. 10112640
kr 189,00 / 1kg -
Catalog No. 10092740
kr 357,30 / 1kg -
Catalog No. 10401841
kr 375,39 / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Bicarbonate, 99.5%-100.0%, (Crystalline powder/ACS Reagent Grade), MP Biomedicals >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction