missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride, 95%, Thermo Scientific™
2844.73 NOK - 9020.00 NOK
Chemical Identifiers
CAS | 270082-22-9 |
---|---|
Molecular Formula | C11H14ClNO2 |
Molecular Weight (g/mol) | 227.69 |
MDL Number | MFCD01861030 |
InChI Key | SWYPWAIQEURSFY-PPHPATTJSA-N |
Synonym | s-2-tetrahydroisoquinoline acetic acid hydrochloride, s-2-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-1,2,3,4-tetrahydro-3-isoquinolineacetic acid hydrochloride, 2-3s-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-2-tetrahydroisoquinoline acetic acid-hcl, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylacetic acid hydrochloride, h-tqa-ohcl, h-tqa-oh.hcl, s-1,2,3,4-tetrahydroisoquinoline-3-acetic acid hydrochloride |
PubChem CID | 2761613 |
IUPAC Name | 2-[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]acetic acid;hydrochloride |
SMILES | [H+].[Cl-].OC(=O)C[C@@H]1CC2=CC=CC=C2CN1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
270082-22-9 | |
227.69 | |
SWYPWAIQEURSFY-PPHPATTJSA-N | |
2761613 | |
[H+].[Cl-].OC(=O)C[C@@H]1CC2=CC=CC=C2CN1 |
C11H14ClNO2 | |
MFCD01861030 | |
s-2-tetrahydroisoquinoline acetic acid hydrochloride, s-2-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-1,2,3,4-tetrahydro-3-isoquinolineacetic acid hydrochloride, 2-3s-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-2-tetrahydroisoquinoline acetic acid-hcl, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylacetic acid hydrochloride, h-tqa-ohcl, h-tqa-oh.hcl, s-1,2,3,4-tetrahydroisoquinoline-3-acetic acid hydrochloride | |
2-[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]acetic acid;hydrochloride |
Specifications
270082-22-9 | |
MFCD01861030 | |
SWYPWAIQEURSFY-PPHPATTJSA-N | |
2-[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]acetic acid;hydrochloride | |
2761613 | |
95% |
C11H14ClNO2 | |
s-2-tetrahydroisoquinoline acetic acid hydrochloride, s-2-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-1,2,3,4-tetrahydro-3-isoquinolineacetic acid hydrochloride, 2-3s-1,2,3,4-tetrahydroisoquinolin-3-yl acetic acid hydrochloride, s-2-tetrahydroisoquinoline acetic acid-hcl, 3s-1,2,3,4-tetrahydroisoquinolin-3-ylacetic acid hydrochloride, h-tqa-ohcl, h-tqa-oh.hcl, s-1,2,3,4-tetrahydroisoquinoline-3-acetic acid hydrochloride | |
[H+].[Cl-].OC(=O)C[C@@H]1CC2=CC=CC=C2CN1 | |
227.69 | |
227.69 | |
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride, 95%, Thermo Scientific™