missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Quantity | 1 g |
Formula Weight | 227.69 |
Percent Purity | 95% |
Chemical Name or Material | (S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
L02W007 | Certificate of Analysis | 06/12/2021 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(S)-1,2,3,4-Tetrahydroisoquinoline-3-acetic acid hydrochloride, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction