missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTEL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1830.00 NOK - 4250.00 NOK
Specifications
Antigen | RTEL1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329515
|
Bio-Techne
NBP3-10036-25UL |
25 μg |
1830.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18315005
|
Bio-Techne
NBP3-10036-100UL |
100 μg |
4250.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RTEL1 Polyclonal specifically detects RTEL1 in Human samples. It is validated for Western Blot.Specifications
RTEL1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, DNA Repair | |
PBS buffer, 2% sucrose | |
51750 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length | |
The immunogen is a synthetic peptide directed towards the middle region of human RTEL1 (NP_057518.1). Peptide sequence HLNQGRPHLSPRPPPTGDPGSQPQWGSGVPRAGKQGQHAVSAYLADARRA | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title