missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTEL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-10036-100UL
This item is not returnable.
View return policy
Description
RTEL1 Polyclonal specifically detects RTEL1 in Human samples. It is validated for Western Blot.Specifications
RTEL1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length | |
The immunogen is a synthetic peptide directed towards the middle region of human RTEL1 (NP_057518.1). Peptide sequence HLNQGRPHLSPRPPPTGDPGSQPQWGSGVPRAGKQGQHAVSAYLADARRA | |
100 μg | |
Cancer, DNA Repair | |
51750 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction