Learn More
Fisherbrand™ Rotator Drum
This Rotator Drum is designed for use with Fisherbrand™ Digital Cel-Gro Tissue Culture Rotator.
3265.00 NOK - 3730.00 NOK
Specifications
Product Type Rotating Drum
View More Specs
Products 3
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
17477632
View Documents
Fisherbrand™
88861240
142 Tubes (17 mm dia.)
3730.00 NOK
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
17487632
View Documents
Fisherbrand™
88861241
76 Tubes (26 mm dia.)
3265.00 NOK
Each
In Stock
to see stock.
Add to basket
17497632
View Documents
Fisherbrand™
88861242
60 Tubes (30 mm dia.)
3265.00 NOK
Each
In Stock
to see stock.
Add to basket
Description
Description
  • For use with Fisherbrand™ Digital Cel-Gro Tissue Culture Rotator
  • Various drum sizes available for rotating different sizes of tubes
Specifications
Specifications
Rotating Drum