Learn More
Fisherbrand™ Rotator Drum
This Rotator Drum is designed for use with Fisherbrand™ Digital Cel-Gro Tissue Culture Rotator.
Brand: Fisherbrand™ 88861242
This item is not returnable.
View return policy
Description
- For use with Fisherbrand™ Digital Cel-Gro Tissue Culture Rotator
- Various drum sizes available for rotating different sizes of tubes
Specifications
60 Tubes (30 mm dia.) | |
Rotating Drum |