missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Antibody (154C1465) - (active/cleaved) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
1530.00 NOK - 4580.00 NOK
Specifications
Antigen | cleaved ROCK1 |
---|---|
Clone | 154C1465 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 21059388) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18064994
|
Bio-Techne
|
0.025 mg |
1530.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788203
|
Novus Biologicals
NB100-56596 |
0.1 mg |
4580.00 NOK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROCK1 Monoclonal specifically detects ROCK1 in Human, Mouse, Rat, Canine, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
cleaved ROCK1 | |
0.5 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway | |
O60259 | |
6093 | |
This antibody was generated by immunizing mice with a synthetic peptide sequence corresponding to amino acids 1114-1126 GNLPESRIEGWLS of human ROCK-1. This sequence is 100% conserved between human, chimpanzee, mouse, rat, dog, and rabbit. This immunogen peptide sequence is adjacent to the caspase-3 cleavage site of ROCK-1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
154C1465 | |
Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 21059388) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Canine, Primate, Rabbit | |
EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1 | |
ROCK1 | |
IgG | |
Protein G purified | |
This mAb recognizes the cleaved C-terminus of ROCK-1. It does not recognize the full-length protein. Since ROCK-1 is cleaved by caspase-3 during apoptosis, this may be used for detection of apoptosis. |
For Research Use Only