Learn More
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths
Reduce operating noise and potential escape of aerosols. Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths are supplied as standard with the bath.
Brand: Grant Instruments™ XAL10
This item is not returnable.
View return policy
Description
- ABS construction
- For use with XUB and XUBA Series Ultrasonic Baths
Specifications
XAB10 Basket for Ultrasonic Bath | |
ABS |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Accessories and Services
Mandatory Accessory
-
Grant Instruments™ XUB Series Digital Ultrasonic Baths DIGITAL ULTRASONIC BATH Product Code: 11450998 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Product Content Correction
Product Title
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths > For Use With: XAB10