Learn More
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths
Reduce operating noise and potential escape of aerosols. Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths are supplied as standard with the bath.
590.00 NOK - 1420.00 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
11431008
|
Grant Instruments™
XAL12 |
XAB12 Basket for Ultrasonic Bath |
1255.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11421008
|
Grant Instruments™
XAL10 |
XAB10 Basket for Ultrasonic Bath | N/A | |||||||
11441008
|
Grant Instruments™
XAL18 |
XAB18 Basket for Ultrasonic Bath |
1290.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11451008
|
Grant Instruments™
XAL25 |
XAB25 Basket for Ultrasonic Bath |
1420.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11411008
|
Grant Instruments™
XAL5 |
XAB5 Basket for Ultrasonic Bath |
928.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11490998
|
Grant Instruments™
XAL1 |
XABA1 Basket for Ultrasonic Bath |
590.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11401008
|
Grant Instruments™
XAL3 |
XABA3 Basket for Ultrasonic Bath |
841.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
Description
- ABS construction
- For use with XUB and XUBA Series Ultrasonic Baths
Specifications
ABS |
Product Title
Grant Instruments™ Replacement ABS Lid for Grant Ultrasonic Baths