Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Brand: Thermo Scientific™ R30166001
4160.02 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
Agglutinating Sera, Haemophilus influenzae Type A | |
H. influenzae | |
Agglutinating Sera |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6110123 | Certificate of Analysis | 23/10/2024 | R30166001 |
3753263 | Certificate of Analysis | 15/03/2024 | R30166001 |
3693056 | Certificate of Analysis | 24/10/2023 | R30166001 |
3631342 | Certificate of Analysis | 13/09/2023 | R30166001 |
3650300 | Certificate of Analysis | 14/08/2023 | R30166001 |