Learn More
BRAND™ Soda-Lime Glass Watch Glasses
Stress-reduced to minimize risk of shattering.
Brand: BRAND™ 150020
345.10 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- Soda-lime glass to DIN 12341
- Ground edges
- Stress-reduced to minimize risk of shattering
Specifications
DIN 12341 | |
Soda Lime Glass | |
60 mm |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
27Hours
17Minutes
59Seconds
49 -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
13Hours
17Minutes
59Seconds
49