Learn More
BINDER™ Reinforced Rack
Material: Stainless Steel; For Use With: KBF 1020, KBF-S 1020
Brand: BINDER™ 8012-0968
This item is not returnable.
View return policy
Description
- Manufactured from stainless steel
- With one set of securing elements (4 pieces)
- Maximum load of 50 kg or 70 kg
Specifications
Rack, reinforced - stainless steel, with fasteners (1 set of 4) | |
Max. 70 kg | |
Rack |
BINDER Chambers KBF 1020, KBF-S 1020 | |
Stainless Steel | |
1 set of 4 fasteners |
Product Content Correction
Product Title
BINDER™ Reinforced Rack > For Use With: KBF 1020