Learn More
Mettler Toledo™ Toledo Potassium Ion Selective Electrode
Brand:  Mettler Toledo™ 51340700
13336.61 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10414831
  • 13675.00 NOK / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Specifications
Polymer Membrane
0°C to 50°C
2 to 12
Product Title
Mettler Toledo™ Toledo Potassium Ion Selective Electrode > Membrane: Polymer