missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-28A/IFN-lambda 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
3710.00 NOK
Specifications
Molecular Weight (g/mol) | 20.5 kDa |
---|---|
Name | IL-28A/IFN-lambda 2 |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived mouse IL-28A/IFN-lambda 2 protein Asp20-Val193,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18136550
|
R&D Systems™
4635-ML-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
3710.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18366367
|
R&D Systems™
4635-ML-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
3710.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.0800-0.800 ng/mL.Specifications
20.5 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
IL-28A/IFN-lambda 2 | |
Mouse myeloma cell line,NS0-derived mouse IL-28A/IFN-lambda 2 protein Asp20-Val193,with a C-terminal 6-His tag | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title