Learn More
R&D Systems™ Recombinant Mouse IL-28A/IFN-lambda 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 4635-ML-025/CF
 View more versions of this product
Product Code. 18136550
  • 3710.00 NOK / 25µg
Explore more special offers
Description
Description
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.0800-0.800 ng/mL.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
IL-28A/IFN-lambda 2
Mouse myeloma cell line,NS0-derived mouse IL-28A/IFN-lambda 2 protein Asp20-Val193,with a C-terminal 6-His tag
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
20.5 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Product Suggestions