missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SNCA / alpha-Synuclein Protein

A DNA sequence encoding the Homo sapiens (Human) SNCA / alpha-Synuclein, was expressed in the hosts and tags indicated.

1644.75 NOK - 19129.63 NOK

Specifications

Accession Number NP_000336.1
Gene ID (Entrez) 6622
Name SNCA / alpha-Synuclein Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16075301
View Documents
enQuireBio™
QP8563-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16015311
View Documents
enQuireBio™
QP8563-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16065301
View Documents
enQuireBio™
QP8563-ec-100ug
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16095301
View Documents
enQuireBio™
QP8563-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16005311
View Documents
enQuireBio™
QP8563-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16085301
View Documents
enQuireBio™
QP8563-ec-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_000336.1
SNCA / alpha-Synuclein Protein
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6622
Research Use Only
Snca
Recombinant Protein
E. coli
MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVATVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGSIAA ATGFVKKDQL GKNEEGAPQE GILEDMPVDP DNEAYEMPSE EGYQDYEPE
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.