missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNCA / alpha-Synuclein Protein
A DNA sequence encoding the Homo sapiens (Human) SNCA / alpha-Synuclein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8563-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_000336.1 | |
SNCA / alpha-Synuclein Protein | |
Research Use Only | |
Snca | |
Recombinant Protein | |
E. coli | |
MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVATVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGSIAA ATGFVKKDQL GKNEEGAPQE GILEDMPVDP DNEAYEMPSE EGYQDYEPE | |
Greater than 90% as determined by SDS-PAGE. |
6622 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction