missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Napsin A Protein
A DNA sequence encoding the Homo sapiens (Human) Napsin A, was expressed in the hosts and tags indicated.
1644.75 NOK - 26695.00 NOK
Specifications
Accession Number | NP_004842.1 |
---|---|
Gene ID (Entrez) | 9476 |
Name | Napsin A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093070
|
enQuireBio™
QP6404-ec-10ug |
10 μg | N/A | ||||||
16033080
|
enQuireBio™
QP6404-ec-50ug |
50 μg | N/A | ||||||
16083070
|
enQuireBio™
QP6404-ec-100ug |
100 μg |
5915.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16013080
|
enQuireBio™
QP6404-ec-200ug |
200 μg | N/A | ||||||
16023080
|
enQuireBio™
QP6404-ec-500ug |
500 μg | N/A | ||||||
16003080
|
enQuireBio™
QP6404-ec-1mg |
1 mg |
26695.00 NOK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_004842.1 | |
Napsin A Protein | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
9476 | |
Research Use Only | |
NAPSA | |
Recombinant Protein | |
KPIFVPLSNY RDVQYFGEIG LGTPPQNFTV AFDTGSSNLW VPSRRCHFFS VPCWLHHRFD PKASSSFQAN GTKFAIQYGT GRVDGILSED KLTIGGIKGA SVIFGEALWE PSLVFAFAHF DGILGLGFPI LSVEGVRPPM DVLVEQGLLD KPVFSFYLNR DPEEPDGGEL VLGGSDPAHY IPPLTFVPVT VPAYWQIHME RVKVGPGLTL CAKGCAAILD TGTSLITGPT EEIRALHAAI GGIPLLAGEY IILCSEIPKL PAVSFLLGGV WFNLTAHDYV IQTTRNGVRL CLSGFQALDV PPPAGPFWIL GDVFLGTYVA VFDRGDMKSS ARVGLARART RGADLGWGET AQAQFPG |