missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Napsin A Protein

A DNA sequence encoding the Homo sapiens (Human) Napsin A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6404-ec-1mg

 View more versions of this product

Product Code. 16003080

  • 26695.00 NOK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004842.1
Napsin A Protein
Research Use Only
NAPSA
Recombinant Protein
E. coli
KPIFVPLSNY RDVQYFGEIG LGTPPQNFTV AFDTGSSNLW VPSRRCHFFS VPCWLHHRFD PKASSSFQAN GTKFAIQYGT GRVDGILSED KLTIGGIKGA SVIFGEALWE PSLVFAFAHF DGILGLGFPI LSVEGVRPPM DVLVEQGLLD KPVFSFYLNR DPEEPDGGEL VLGGSDPAHY IPPLTFVPVT VPAYWQIHME RVKVGPGLTL CAKGCAAILD TGTSLITGPT EEIRALHAAI GGIPLLAGEY IILCSEIPKL PAVSFLLGGV WFNLTAHDYV IQTTRNGVRL CLSGFQALDV PPPAGPFWIL GDVFLGTYVA VFDRGDMKSS ARVGLARART RGADLGWGET AQAQFPG
Greater than 90% as determined by SDS-PAGE.
9476
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers