missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-11 / interleukin 11 Protein

A DNA sequence encoding the Human IL-11 / interleukin 11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10319-ec-250ug

 View more versions of this product

Product Code. 15933488

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000632.1
IL-11 / interleukin 11 Protein
Research Use Only
IL11
The ED50 as determined by a cell proliferation assay using murine B9-11 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
3589
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL
Greater than 95% as determined by SDS-PAGE and HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers