missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-11 / interleukin 11 Protein

A DNA sequence encoding the Human IL-11 / interleukin 11, was expressed in the hosts and tags indicated.

1085.75 NOK - 35292.25 NOK

Specifications

Accession Number NP_000632.1
Gene ID (Entrez) 3589
Name IL-11 / interleukin 11 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15943488
View Documents Promotion Details
enQuireBio™
QP10319-ec-2ug
2 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15923488
View Documents Promotion Details
enQuireBio™
QP10319-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15913488
View Documents Promotion Details
enQuireBio™
QP10319-ec-100ug
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15933488
View Documents Promotion Details
enQuireBio™
QP10319-ec-250ug
250 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_000632.1
IL-11 / interleukin 11 Protein
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL
3589
Research Use Only
IL11
The ED50 as determined by a cell proliferation assay using murine B9-11 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Human
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documents

Documents

Product Certifications