Learn More
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
410.00 NOK - 463.00 NOK
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
10341190
|
Lenz Laborglasintrumente
05272014 |
Socket and Cone NS 29/32 |
463.00 NOK
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
11516824
|
Lenz Laborglasintrumente
05272012 |
Socket and Cone NS 19/26 |
410.00 NOK
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
11526824
|
Lenz Laborglasintrumente
05272013 |
Socket and Cone NS 24/29 |
463.00 NOK
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
Description
- Receiver adapters acc. to DIN 12594
- Straight
- With NS socket and NS cone
- With drain tip
- Vacuum connection: thread GL18
- Made of DURAN™ tubing
Specifications
Duran Glass |
Product Title
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread