Learn More
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05272012
 View more versions of this product
Product Code. 11516824
  • 410.00 NOK / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Receiver adapters acc. to DIN 12594
  • Straight
  • With NS socket and NS cone
  • With drain tip
  • Vacuum connection: thread GL18
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Glass
Socket and Cone NS 19/26
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Receiver Adapters, Straight, With GL Thread > Socket and cone NS 19/26