Learn More
Invitrogen™ ReadyCount™ Stains
The ReadyCount stains are benchtop-stable fluorescent reagents for the Countess II FL and 3 FL automated cell counters. They are ready-to-use to enable fast and easy automated counting of nucleated cells or isolated nuclei.
Brand:  Invitrogen™ A49904
 View more versions of this product
Product Code. 17108865
  • 1168.00 NOK / Pack of 100
Explore more special offers
Description
Show More
Specifications
Specifications
Nuclear Stain
Countess II FL >1.0.249; Countess 3 FL ≥1.0.296.782
Blue
100 Assays
ReadyCount
Countess II FL, Countess 3 FL
ReadyCount™ Blue Nuclear Stain, 1 x 1 mL vial
Stable at room temperature.
Ambient
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3123549 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberA49904
Lot Number2680206 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog NumberA49904
Lot Number2615838 Certificate TypeCertificate of Analysis Date20/03/2023 Catalog NumberA49904
Lot Number2518904 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA49904
Lot Number2495432 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog NumberA49904
Product Title
Invitrogen™ ReadyCount™ Stains > Blue Nuclear Stain