Learn More
IKA™ RCT Digital Magnetic Stirring Hotplate
RCT digital stirrer has a round top made of aluminum alloy, stirs volumes of up to 20 L (H2O), and runs at speeds ranging from 50 to1500 rpm.
15010.00 NOK
Product Code | Brand | Plug Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Type | Price | Quantity & Availability | ||||||
15673687
![]() |
IKA™
0025004601 |
EU |
15010.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
Description
- Strong motor for a high range of speeds
- Additional temperature control mode for faster heating
- Exact temperature and speed setting via digital display
- Set safety temperature limit displayed digitally
- Hot top indicator
- Digital error code display
- Enclosed assembly guarantees long service life
Product Title
IKA™ RCT Digital Magnetic Stirring Hotplate