Learn More
IKA™ RCT Digital Magnetic Stirring Hotplate
RCT digital stirrer has a round top made of aluminum alloy, stirs volumes of up to 20 L (H2O), and runs at speeds ranging from 50 to1500 rpm.
Brand: IKA™ 0025004601
11051.43 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
- Strong motor for a high range of speeds
- Additional temperature control mode for faster heating
- Exact temperature and speed setting via digital display
- Set safety temperature limit displayed digitally
- Hot top indicator
- Digital error code display
- Enclosed assembly guarantees long service life
Specifications
EU |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%