Learn More
Rad51D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11126-1-AP-20UL
 View more versions of this product
Product Code. 16847993
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
Rad52 family members (Rad50, Rad51B/C/D, Rad52, Rad54, MRE11) mediate DNA double-strand break repair (DSBR) for DNA damage that otherwise could cause cell death, mutation or neoplastic transformation. Rad51 (RECA, BRCC5) interacts with BRCA1 and BRCA2 to influence subcellular localization and cellular response to DNA damage. BRCA2 inactivation may be a key event leading to genomic instability and tumorigenesis from deregulation of Rad51. Rad52 forms a heptameric ring that binds single-stranded DNA ends and catalyzes DNA-DNA interaction necessary for the annealing of complementary strands. Rad52 can interact with Rad51. Rad54A of the DEAD-like helicase superfamily binds to double-strand DNA and induces a DNA topological change, which is thought to facilitate homologous DNA pairing and stimulate DNA recombination. Rad54B of the DEAD-like helicase superfamily binds to double-stranded DNA and displays ATPase activity in the presence of DNA. Rad54B is abundant in testis and spleen, and mutations of this gene occur in primary lymphoma and colon cancer
Specifications
Specifications
Rad51D
Polyclonal
Unconjugated
RAD51D
HsTRAD, R51H3, RAD51 homolog D, RAD51 like 3 (S. cerevisiae), RAD51 like protein 3, Rad51D, RAD51L3, Trad
Rabbit
Antigen Affinity Chromatography
RUO
19364, 303375, 5892
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O55230, O75771
RAD51D
Rad51D Fusion Protein Ag1595
20 μL
Primary
Human, Mouse, Rat