Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Ideally suited for use with volumetric flasks.
Brand:  Quickfit TRF691
795.11 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 12421788
  • 872.00 NOK / Pack of 10
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Description
Ideally suited for use with volumetric flasks.
  • Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
  • Hexagonal head prevents stopper from rolling and provides good grip
  • Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
  • Hollow-blown to reduce weight
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Volumetric flasks
10/19
Glass
Hollow Stopper
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    15
    Hours
    8
    Minutes
    39
    Seconds
    13
Product Content Correction
Product Title
Quickfit™ Hexagonal Hollow-blown Glass Stopper > Cone Size: 10/19