Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 01400010
 View more versions of this product
Product Code. 10495251
  • 166.00 NOK / Pack of 10
Only null left
Estimated Shipment: 28-07-2025
Description
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hollow
Thread Size: 10/19 Cone NS
Hollow Stopper
10/Pk.
Borosilicate Glass
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate > Thread Size: 10/19 Cone NS