missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCRL Mouse anti-Human, Rabbit, Clone: 1G7A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68087-1-IG-150UL
This item is not returnable.
View return policy
Description
PYCRL Monoclonal antibody apecifically detetects PYCRL antigen in Human, Rabbit Samples. It is validated for Western Blot, ELISA.Specifications
PYCRL | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
PYCRL | |
PYCRL fusion protein Ag31938 | |
150 μL | |
65263 | |
Store at -20°C. Stable for one year after shipment. | |
IgG1 |
Western Blot, ELISA | |
1G7A3 | |
Unconjugated | |
BC007993 | |
Mouse | |
Protein G purification | |
Primary | |
Human, Rabbit | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction