missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCRL Mouse anti-Human, Rabbit, Clone: 1G7A3, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | PYCRL |
---|---|
Clone | 1G7A3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17609776
|
Proteintech
68087-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17699766
|
Proteintech
68087-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PYCRL Monoclonal antibody apecifically detetects PYCRL antigen in Human, Rabbit Samples. It is validated for Western Blot, ELISA.Specifications
PYCRL | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Rabbit | |
BC007993 | |
PYCRL | |
IgG1 | |
Protein G purification |
1G7A3 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
65263 | |
PYCRL fusion protein Ag31938 | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title