Learn More
BRAND™ PTFE Watch Glass
PTFE construction. Ideal for covering PTFE beakers.
Brand: BRAND™ 150915
318.79 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- PTFE construction
- Excellent chemical resistance
- Ideal for covering PTFE beakers
Specifications
Flat | |
Round | |
Covering PTFE Beakers |
PTFE | |
Watch Glass | |
100 mm |
Product Content Correction
Product Title
BRAND™ PTFE Watch Glass > Diameter: 100mm