Learn More
BRAND™ PTFE Watch Glass
PTFE construction. Ideal for covering PTFE beakers.
253.00 NOK - 327.00 NOK
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
10718361
|
BRAND™
150912 |
80 mm |
253.00 NOK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
10485811
|
BRAND™
150915 |
100 mm |
327.00 NOK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||||
Description
- PTFE construction
- Excellent chemical resistance
- Ideal for covering PTFE beakers
Specifications
Flat | |
PTFE | |
Round | |
Watch Glass |
Product Title
BRAND™ PTFE Watch Glass