Learn More
Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector
Made from Kynar. Ark-Plas Products™ Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector is ideal for connecting tubes of various diameter. The connector offers easy fitting into tubing or hoses without the need for tools.
183.00 NOK
Specifications
Product Type | Straight reducing connector |
---|---|
Material | Fluororesin |
Color | Natural |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
12313850
|
Ark-Plas Products™
MAD80-DF0 |
5/32 x 1/16 in. |
183.00 NOK
Pack of 10 |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
12323850
|
Ark-Plas Products™
MAD60-DF0 |
3/16 x 1/16 in. |
183.00 NOK
Pack of 10 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
Description
- Made of Kynar
- Economical straight barbed
- Easily connect fluid paths without the use of tools
Specifications
Straight reducing connector | |
Fluororesin | |
Natural |